Learn More
Biomatik Corporation Recombinant Human ATP synthase subunit delta, mitochondrial (ATP5D), 100UG
Supplier:  Biomatik Corporation RPC20008
Encompass_Preferred
Recombinant Human ATP synthase subunit delta, mitochondrial (ATP5D) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ATP5D. Target Synonyms: F-ATPase delta subunit. Accession Number: P30049. Expression Region: 23~168aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 31kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4537