Learn More
Biomatik Corporation Recombinant Bacteroides fragilis Chaperone protein htpG (htpG), partial, 100UG
Supplier:  Biomatik Corporation RPC23135
Encompass_Preferred
Recombinant Bacteroides fragilis Chaperone protein htpG (htpG), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bacteroides fragilis (strain ATCC 25285 / DSM 2151 / JCM 11019 / NCTC 9343). Target Name: htpG. Target Synonyms: Heat shock protein HtpGHigh temperature protein G. Accession Number: Q5LCH4. Expression Region: 1~326aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 57kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5240