Learn More
Biomatik Corporation Recombinant Mouse Complement component receptor 1-like protein (Cr1l), partial, 100UG
Supplier:  Biomatik Corporation RPC23152
Encompass_Preferred
Recombinant Mouse Complement component receptor 1-like protein (Cr1l), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Cr1l. Target Synonyms: Complement regulatory protein CrryProtein p65. Accession Number: Q64735. Expression Region: 41~405aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 44.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5242