Learn More
Biomatik Corporation Recombinant Human HERV-K_7p22.1 provirus ancestral Env polyprotein (ERVK-6), partial, 100UG
Supplier:  Biomatik Corporation RPC23170
Encompass_Preferred
Recombinant Human Endogenous retrovirus group K member 6 Env polyprotein (ERVK-6), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ERVK-6. Target Synonyms: EnvK2 proteinEnvelope polyproteinHERV-K(C7) envelope proteinHERV-K(HML-2.HOM) envelope proteinHERV-K108 envelope proteinHERV-K_7p22.1 provirus ancestral Env polyprotein. Accession Number: Q69384. Expression Region: 90~632aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 65.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5245