Learn More
Biomatik Corporation Recombinant Streptococcus pyogenes serotype M6 50S ribosomal protein L7/L12 (rplL), 100UG
Supplier:  Biomatik Corporation RPC23179
Encompass_Preferred
Recombinant Streptococcus pyogenes serotype M6 50S ribosomal protein L7/L12 (rplL) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Streptococcus pyogenes serotype M6 (strain ATCC BAA-946 / MGAS10394). Target Name: rplL. Target Synonyms: rplL; M6_Spy0814; 50S ribosomal protein L7/L12. Accession Number: Q5XCB4. Expression Region: 1~121aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 32.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5246