Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Clumping factor AclfA), partial, 100UG
Supplier:  Biomatik Corporation RPC23222
Encompass_Preferred
Recombinant Staphylococcus aureus Clumping factor A(clfA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Staphylococcus aureus (strain MSSA476). Target Name: clfA. Target Synonyms: Fibrinogen receptor AFibrinogen-binding protein A. Accession Number: Q6GB45. Expression Region: 228~558aa. Tag Info: Tag-Free. Theoretical MW: 36.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5249