Learn More
Biomatik Corporation Recombinant Zaire ebolavirus Minor nucleoprotein VP30 (VP30), 100UG
Supplier:  Biomatik Corporation RPC23232
Encompass_Preferred
Recombinant Zaire ebolavirus Minor nucleoprotein VP30 (VP30) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Zaire ebolavirus (strain Kikwit-95) (ZEBOV) (Zaire Ebola virus). Target Name: VP30. Target Synonyms: Transcription activator VP30. Accession Number: Q77DJ5. Expression Region: 1~288aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 38kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5251