Learn More
Biomatik Corporation Recombinant Human Inter-alpha-trypsin inhibitor heavy chain H5 (ITIH5)Partial, 100UG
Supplier:  Biomatik Corporation RPC23253
Encompass_Preferred
Recombinant Human Inter-alpha-trypsin inhibitor heavy chain H5 (ITIH5), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ITIH5. Target Synonyms: ITIH5; KIAA1953; PP14776; UNQ311/PRO354; Inter-alpha-trypsin inhibitor heavy chain H5; ITI heavy chain H5; ITI-HC5; Inter-alpha-inhibitor heavy chain 5. Accession Number: Q86UX2. Expression Region: 35~161aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 30.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5256