Learn More
Biomatik Corporation Recombinant Pig Interleukin-4 receptor subunit alpha (IL4R), partial, 100UG
Supplier:  Biomatik Corporation RPC23263
Encompass_Preferred
Recombinant Pig Interleukin-4 receptor subunit alpha (IL4R), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Pig (Sus scrofa; Porcine). Target Name: IL4R. Target Synonyms: CD_antigen: CD124. Accession Number: Q863Z5. Expression Region: 33~240aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 28.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5257