Learn More
Biomatik Corporation Recombinant Human Sprouty-related, EVH1 domain-containing protein 1 (SPRED1), 100UG
Supplier:  Biomatik Corporation RPC23269
Encompass_Preferred
Recombinant Human Sprouty-related, EVH1 domain-containing protein 1 (SPRED1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SPRED1. Target Synonyms: EVH1 domain-containing protein 1; EVH1/Sprouty domain containing protein; FLJ33903; hSpred 1; hSpred1; NFLS; PPP1R147; protein phosphatase 1 regulatory subunit 147; SPRE1_HUMAN; SPRED 1; Spred-1; spred1. Accession Number: Q7Z699. Expression Region: 2~444aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 70.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5258