Learn More
Biomatik Corporation Recombinant Ambrosia artemisiifolia Pectate lyase 5, 100UG
Supplier:  Biomatik Corporation RPC20116
Encompass_Preferred
Recombinant Ambrosia artemisiifolia Pectate lyase 5 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Ambrosia artemisiifolia (Short ragweed). Target Name: Ambrosia artemisiifolia Pectate lyase 5. Target Synonyms: Pectate lyase 5; EC 4.2.2.2; Antigen Amb a I; Antigen E; AgE; Pollen allergen Amb a 1.1; allergen Amb a 1.1. Accession Number: P27759. Expression Region: 26~396aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 55.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4602