Learn More
Biomatik Corporation Recombinant Macaca fascicularis Frataxin, mitochondrial (FXN), 100UG
Supplier:  Biomatik Corporation RPC23287
Encompass_Preferred
Recombinant Macaca fascicularis Frataxin, mitochondrial (FXN) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Macaca fascicularis (Crab-eating macaque) (Cynomolgus monkey). Target Name: FXN. Target Synonyms: FXN; FRDA1; QnpA-13971; Frataxin; mitochondrial; Fxn; EC 1.16.3.1; Cleaved into: Frataxin intermediate form; Frataxin mature form. Accession Number: Q8HXX9. Expression Region: 81~210aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 18.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5260