Learn More
Biomatik Corporation Recombinant Vibrio vulnificus Fe/S biogenesis protein NfuA (nfuA), 100UG
Supplier:  Biomatik Corporation RPC23297
Encompass_Preferred
Recombinant Vibrio vulnificus Fe/S biogenesis protein NfuA (nfuA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Vibrio vulnificus (strain CMCP6). Target Name: nfuA. Target Synonyms: nfuA; VV1_0864; Fe/S biogenesis protein NfuA. Accession Number: Q8DDU2. Expression Region: 1~194aa. Tag Info: Tag-Free. Theoretical MW: 21kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5261