Learn More
Biomatik Corporation Recombinant Buchnera aphidicola subsp. Schizaphis graminum Porin-like protein BUsg_347 (BUsg_347), 100UG
Supplier:  Biomatik Corporation RPC23301
Encompass_Preferred
Recombinant Buchnera aphidicola subsp. Schizaphis graminum Porin-like protein BUsg_347 (BUsg_347) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Buchnera aphidicola subsp. Schizaphis graminum (strain Sg). Target Name: BUsg_347. Target Synonyms: BUsg_347; Porin-like protein BUsg_347. Accession Number: Q8K9I8. Expression Region: 24~369aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 59kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5262