Learn More
Biomatik Corporation Recombinant Human herpesvirus 1 Envelope glycoprotein L (gL), 100UG
Catalog No. 501505264
Encompass_Preferred
Recombinant Human herpesvirus 1 Envelope glycoprotein L (gL) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human herpesvirus 1 (strain KOS) (HHV-1) (Human herpes simplex virus 1). Target Name: gL. Target Synonyms: gL; UL1Envelope glycoprotein L; gL. Accession Number: Q96912. Expression Region: 23~224aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 42.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5264 Supplier Biomatik Corporation Supplier No. RPC23347
Recombinant Human herpesvirus 1 Envelope glycoprotein L (gL) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human herpesvirus 1 (strain KOS) (HHV-1) (Human herpes simplex virus 1). Target Name: gL. Target Synonyms: gL; UL1Envelope glycoprotein L; gL. Accession Number: Q96912. Expression Region: 23~224aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 42.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.