Learn More
Biomatik Corporation Recombinant Aspergillus kawachii Probable endo-beta-1,4-glucanase D (eglD), 100UG
Supplier:  Biomatik Corporation RPC23360
Encompass_Preferred
Recombinant Aspergillus kawachii Probable endo-beta-1, 4-glucanase D (eglD) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Aspergillus kawachii (strain NBRC 4308) (White koji mold) (Aspergillus awamori var. kawachi). Target Name: eglD. Target Synonyms: Carboxymethylcellulase DCellulase 61ACellulase Dcel61A. Accession Number: Q96WQ9. Expression Region: 21~408aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 45.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5265