Learn More
Biomatik Corporation Recombinant Human Selenoprotein M (SELM), 100UG
Catalog No. 501505266
Encompass_Preferred
Recombinant Human Selenoprotein M (SELENOM) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SELENOM. Target Synonyms: SELENOM; SELM; Selenoprotein M; SelM. Accession Number: Q8WWX9. Expression Region: 24~145aa. Tag Info: Tag-Free. Theoretical MW: 13.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5266 Supplier Biomatik Corporation Supplier No. RPC23370
Recombinant Human Selenoprotein M (SELENOM) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SELENOM. Target Synonyms: SELENOM; SELM; Selenoprotein M; SelM. Accession Number: Q8WWX9. Expression Region: 24~145aa. Tag Info: Tag-Free. Theoretical MW: 13.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.