Learn More
Biomatik Corporation Recombinant Carpinus betulus Major pollen allergen Car b 1 isoforms 1A and 1B, 100UG
Supplier:  Biomatik Corporation RPC20117
Encompass_Preferred
Recombinant Carpinus betulus Major pollen allergen Car b 1 isoforms 1A and 1B is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Carpinus betulus (European hornbeam) (Carpinus caucasica). Target Name: Carpinus betulus Major pollen allergen Car b 1 isoforms 1A and 1B. Target Synonyms: Major pollen allergen Car b 1 isoforms 1A and 1B; Allergen Car b I; allergen Car b 1. Accession Number: P38949. Expression Region: 2~160aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 21.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4603