Learn More
Biomatik Corporation Recombinant Mouse Secretoglobin family 3A member 2 (Scgb3a2), 100UG
Supplier:  Biomatik Corporation RPC23418
Encompass_Preferred
Recombinant Mouse Secretoglobin family 3A member 2 (Scgb3a2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Scgb3a2. Target Synonyms: Pneumo secretory protein 1. Accession Number: Q920H1. Expression Region: 22~139aa. Tag Info: N-Terminal 6Xhis-Gst-Tagged. Theoretical MW: 43.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5273