Learn More
Biomatik Corporation Recombinant Human Trimethylguanosine synthase (TGS1), partial, 100UG
Supplier:  Biomatik Corporation RPC23438
Encompass_Preferred
Recombinant Human Trimethylguanosine synthase (TGS1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TGS1. Target Synonyms: CLL-associated antigen KW-2; Cap-specific guanine-N2 methyltransferaseHepatocellular carcinoma-associated antigen 137Nuclear receptor coactivator 6-interacting protein; PRIP-interacting protein with methyltransferase motif; PIMT; PIPMT. Accession Number: Q96RS0. Expression Region: 713~853aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 31.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5274