Learn More
Biomatik Corporation Recombinant Pig Interleukin-5 (IL5), 100UG
Supplier:  Biomatik Corporation RPC23549
Encompass_Preferred
Recombinant Pig Interleukin-5 (IL5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Pig (Sus scrofa; Porcine). Target Name: IL5. Target Synonyms: Eosinophil differentiation factorT-cell replacing factor. Accession Number: Q9MYM5. Expression Region: 22~134aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 17kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5283