Learn More
Biomatik Corporation Recombinant Human Interleukin-26 (IL26), 100UG
Catalog No. 501505286
Encompass_Preferred
Recombinant Human Interleukin-26 (IL26) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IL26. Target Synonyms: Protein AK155AK155. Accession Number: Q9NPH9. Expression Region: 22~171aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 31.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5286 Supplier Biomatik Corporation Supplier No. RPC23615
Recombinant Human Interleukin-26 (IL26) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IL26. Target Synonyms: Protein AK155AK155. Accession Number: Q9NPH9. Expression Region: 22~171aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 31.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.