Learn More
Biomatik Corporation Recombinant Mouse C-type lectin domain family 4 member A (Clec4a), partial, 100UG
Supplier:  Biomatik Corporation RPC23625
Encompass_Preferred
Recombinant Mouse C-type lectin domain family 4 member A (Clec4a), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Clec4a. Target Synonyms: C-type lectin superfamily member 6Dendritic cell immunoreceptor. Accession Number: Q9QZ15. Expression Region: 70~238aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 39.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5288