Learn More
Biomatik Corporation Recombinant Salmonella typhimurium Protein prgI (prgI), 100UG
Supplier:  Biomatik Corporation RPC20119
Encompass_Preferred
Recombinant Salmonella typhimurium Protein prgI (prgI) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Salmonella typhimurium (strain LT2 / SGSC1412 / ATCC 700720). Target Name: prgI. Target Synonyms: prgI; STM2873; Protein PrgI. Accession Number: P41784. Expression Region: 1~80aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 24.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4605