Learn More
Biomatik Corporation Recombinant Influenza A virus Polymerase acidic protein (PA), partial, 100UG
Catalog No. 501505291
Encompass_Preferred
Recombinant Influenza A virus Polymerase acidic protein (PA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Influenza A virus (strain A/x-31 H3N2). Target Name: PA. Target Synonyms: RNA-directed RNA polymerase subunit P2. Accession Number: Q9IQ47. Expression Region: 124~247aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 18.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5291 Supplier Biomatik Corporation Supplier No. RPC23675
Recombinant Influenza A virus Polymerase acidic protein (PA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Influenza A virus (strain A/x-31 H3N2). Target Name: PA. Target Synonyms: RNA-directed RNA polymerase subunit P2. Accession Number: Q9IQ47. Expression Region: 124~247aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 18.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.