Learn More
Biomatik Corporation Recombinant Human Hydroxyacid oxidase 2 (HAO2), 100UG
Supplier:  Biomatik Corporation RPC23680
Encompass_Preferred
Recombinant Human Hydroxyacid oxidase 2 (HAO2) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HAO2. Target Synonyms: (S)-2-hydroxy-acid oxidase, peroxisomalCell growth-inhibiting gene 16 proteinLong chain alpha-hydroxy acid oxidaseLong-chain L-2-hydroxy acid oxidase. Accession Number: Q9NYQ3. Expression Region: 2~351aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 54.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5293