Learn More
Biomatik Corporation Recombinant Human tRNA methyltransferase 112 homolog (TRMT112), 100UG
Supplier:  Biomatik Corporation RPC23688
Encompass_Preferred
Recombinant Human Multifunctional methyltransferase subunit TRM112-like protein (TRMT112) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TRMT112. Target Synonyms: tRNA methyltransferase 112 homolog. Accession Number: Q9UI30. Expression Region: 1~125aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 30.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5294