Learn More
Biomatik Corporation Recombinant Mouse Hydroxyacid oxidase 1 (Hao1), 100UG
Catalog No. 501505296
Encompass_Preferred
Recombinant Mouse 2-Hydroxyacid oxidase 1 (Hao1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Hao1. Target Synonyms: HAOX1Glycolate oxidaseGOX. Accession Number: Q9WU19. Expression Region: 1~370aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 45kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5296 Supplier Biomatik Corporation Supplier No. RPC23764
Recombinant Mouse 2-Hydroxyacid oxidase 1 (Hao1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Hao1. Target Synonyms: HAOX1Glycolate oxidaseGOX. Accession Number: Q9WU19. Expression Region: 1~370aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 45kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.