Learn More
Biomatik Corporation Recombinant Cryptomeria japonica Polygalacturonase, 100UG
Supplier:  Biomatik Corporation RPC20121
Encompass_Preferred
Recombinant Cryptomeria japonica Polygalacturonase is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Cryptomeria japonica (Japanese cedar) (Cupressus japonica). Target Name: Cryptomeria japonica Polygalacturonase. Target Synonyms: Polygalacturonase; PG; EC 3.2.1.15; Allergen Cry j II; Major pollen allergen Cry j 2; Pectinase; allergen Cry j 2. Accession Number: P43212. Expression Region: 46~433aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 58.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4606