Learn More
Biomatik Corporation Recombinant Macaca mulatta Interleukin-10 (IL10), 100UG
Supplier:  Biomatik Corporation RPC23829
Encompass_Preferred
Recombinant Macaca mulatta Interleukin-10 (IL10) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Macaca mulatta (Rhesus macaque). Target Name: IL10. Target Synonyms: Cytokine synthesis inhibitory factor. Accession Number: P51496. Expression Region: 19~178aa. Tag Info: C-Terminal 10Xhis-Tagged. Theoretical MW: 20.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5300