Learn More
Biomatik Corporation Recombinant Escherichia coli Transcriptional regulatory protein phoP (phoP), 100UG
Supplier:  Biomatik Corporation RPC23865
Encompass_Preferred
Recombinant Escherichia coli Transcriptional regulatory protein phoP (phoP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: phoP. Target Synonyms: phoP; b1130; JW1116; Transcriptional regulatory protein PhoP. Accession Number: P23836. Expression Region: 1~223aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 29.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5305