Learn More
Biomatik Corporation Recombinant Mouse Transcription factor MafK (Mafk), 100UG
Supplier:  Biomatik Corporation RPC23879
Encompass_Preferred
Recombinant Mouse Transcription factor MafK (Mafk) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Mafk. Target Synonyms: Erythroid transcription factor NF-E2 p18 subunit. Accession Number: Q61827. Expression Region: 1~156aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 21.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5306