Learn More
Biomatik Corporation Recombinant Rat Sclerostin domain-containing protein 1 (Sostdc1), 100UG
Supplier:  Biomatik Corporation RPC23881
Encompass_Preferred
Recombinant Rat Sclerostin domain-containing protein 1 (Sostdc1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Sostdc1. Target Synonyms: Uterine sensitization-associated gene 1 protein. Accession Number: Q642G2. Expression Region: 24~206aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 24.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5307