Learn More
Biomatik Corporation Recombinant Human Sialic acid-binding Ig-like lectin 15 (SIGLEC15), partial, 100UG
Supplier:  Biomatik Corporation RPC23884
Encompass_Preferred
Recombinant Human Sialic acid-binding Ig-like lectin 15 (SIGLEC15), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SIGLEC15. Target Synonyms: CD33 antigen-like 3. Accession Number: Q6ZMC9. Expression Region: 20~263aa. Tag Info: N-Terminal 6Xhis-Myc-Tagged. Theoretical MW: 30.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5308