Learn More
Biomatik Corporation Recombinant Onchocerca volvulus OV-16 antigen (OV16), 100UG
Catalog No. 501504607
Encompass_Preferred
Recombinant Onchocerca volvulus OV-16 antigen (OV16) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Onchocerca volvulus. Target Name: OV16. Target Synonyms: OV16; OV-16 antigen. Accession Number: P31729. Expression Region: 17~197aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 24kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4607 Supplier Biomatik Corporation Supplier No. RPC20123
Recombinant Onchocerca volvulus OV-16 antigen (OV16) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Onchocerca volvulus. Target Name: OV16. Target Synonyms: OV16; OV-16 antigen. Accession Number: P31729. Expression Region: 17~197aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 24kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.