Learn More
Biomatik Corporation Recombinant Zaire ebolavirus Polymerase cofactor VP35 (VP35), 100UG
Catalog No. 501505309
Encompass_Preferred
Recombinant Zaire ebolavirus Polymerase cofactor VP35 (VP35) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Zaire ebolavirus (strain Kikwit-95) (ZEBOV) (Zaire Ebola virus). Target Name: VP35. Target Synonyms: VP35Polymerase cofactor VP35; Ebola VP35; eVP35. Accession Number: Q6V1Q9. Expression Region: 1~340aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 41.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5309 Supplier Biomatik Corporation Supplier No. RPC23885
Recombinant Zaire ebolavirus Polymerase cofactor VP35 (VP35) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Zaire ebolavirus (strain Kikwit-95) (ZEBOV) (Zaire Ebola virus). Target Name: VP35. Target Synonyms: VP35Polymerase cofactor VP35; Ebola VP35; eVP35. Accession Number: Q6V1Q9. Expression Region: 1~340aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 41.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.