Learn More
Biomatik Corporation Recombinant Human Glucose-6-phosphate isomerase (GPI), partial, 100UG
Supplier:  Biomatik Corporation RPC23913
Encompass_Preferred
Recombinant Human Glucose-6-phosphate isomerase (GPI), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GPI. Target Synonyms: Autocrine motility factor; AMFNeuroleukin; NLKPhosphoglucose isomerase; PGIPhosphohexose isomerase; PHISperm antigen 36; SA-36. Accession Number: P06744. Expression Region: 2~554aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 66.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5313