Recombinant Human 40S ribosomal protein S13 (RPS13), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RPS13. Target Synonyms: 2700063M04Rik; 40S ribosomal protein S13; MGC102403; MGC118043; Ribosomal protein S13; RPS13; RS13_HUMAN. Accession Number: P62277. Expression Region: 3~151aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 44kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.