Learn More
Biomatik Corporation Recombinant Human Collagen alpha-2 (IV) chain (COL4A2), partial, 100UG
Catalog No. 501505326
Encompass_Preferred
Recombinant Human Collagen alpha-2 (IV) chain (COL4A2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: COL4A2. Target Synonyms: Canstatin; CO4A2_HUMAN; COL4A 2; Col4a2; Collagen alpha 2(IV) chain; Collagen; type IV; alpha 2. Accession Number: P08572. Expression Region: 1493~1712aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 28.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5326 Supplier Biomatik Corporation Supplier No. RPC24283
Recombinant Human Collagen alpha-2 (IV) chain (COL4A2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: COL4A2. Target Synonyms: Canstatin; CO4A2_HUMAN; COL4A 2; Col4a2; Collagen alpha 2(IV) chain; Collagen; type IV; alpha 2. Accession Number: P08572. Expression Region: 1493~1712aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 28.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.