Learn More
Biomatik Corporation Recombinant Human HLA-C protein (HLA-C), partial, 100UG
Supplier:  Biomatik Corporation RPC24351
Encompass_Preferred
Recombinant Human HLA class I histocompatibility antigen, C alpha chain (HLA-C), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HLA-C. Accession Number: O78179. Expression Region: 25~308aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 36.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5328