Learn More
Biomatik Corporation Recombinant Centruroides suffusus suffusus Beta-mammal toxin Css4, partial, 100UG
Supplier:  Biomatik Corporation RPC20125
Encompass_Preferred
Recombinant Centruroides suffusus Beta-mammal toxin Css4 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Centruroides suffusus suffusus (Mexican scorpion). Target Name: Centruroides suffusus Beta-mammal toxin Css4. Target Synonyms: Beta-mammal toxin Css4; Css IV; CssIV. Accession Number: P60266. Expression Region: 20~85aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 23.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4609