Learn More
Biomatik Corporation Recombinant Human Cathepsin F protein (CTSF), partial, 100UG
Catalog No. 501505329
Encompass_Preferred
Recombinant Human Cathepsin F (CTSF), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CTSF. Target Synonyms: AI481912; CATF_HUMAN; Cathepsin F; CathepsinF; CATSF; CLN13; Ctsf; EC 3.4.22.41. Accession Number: Q9UBX1. Expression Region: 273~484aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5329 Supplier Biomatik Corporation Supplier No. RPC24354
Recombinant Human Cathepsin F (CTSF), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CTSF. Target Synonyms: AI481912; CATF_HUMAN; Cathepsin F; CathepsinF; CATSF; CLN13; Ctsf; EC 3.4.22.41. Accession Number: Q9UBX1. Expression Region: 273~484aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.