Learn More
Biomatik Corporation Recombinant Escherichia coli Anaerobic ribonucleoside-triphosphate reductase (nrdD), 100UG
Supplier:  Biomatik Corporation RPC24371
Encompass_Preferred
Recombinant Escherichia coli Anaerobic ribonucleoside-triphosphate reductase (nrdD) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: nrdD. Target Synonyms: nrdD; b4238; JW4197; Anaerobic ribonucleoside-triphosphate reductase; EC 1.1.98.6; Class III ribonucleoside-triphosphate reductase. Accession Number: P28903. Expression Region: 1~712aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 84kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5332