Learn More
Biomatik Corporation Recombinant Streptococcus pyogenes serotype M1 Adenine phosphoribosyltransferase (apt), 100UG
Supplier:  Biomatik Corporation RPC24508
Encompass_Preferred
Recombinant Streptococcus pyogenes serotype M1 Adenine phosphoribosyltransferase (apt) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Streptococcus pyogenes serotype M1. Target Name: apt. Target Synonyms: apt; SPy_0927; M5005_Spy0728Adenine phosphoribosyltransferase; APRT; EC 2.4.2.7. Accession Number: P63546. Expression Region: 1~172aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 22.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5337