Learn More
Biomatik Corporation RecombinantMouseAquaporin-4 (Aqp4), partial, 100UG
Supplier:  Biomatik Corporation RPC24509
Encompass_Preferred
Recombinant MouseAquaporin-4 (Aqp4), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Aqp4. Target Synonyms: Mercurial-insensitive water channel. Accession Number: P55088. Expression Region: 253~323aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 9.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5338