Learn More
Biomatik Corporation Recombinant Human Renin receptor (ATP6AP2), 100UG
Supplier:  Biomatik Corporation RPC20009
Encompass_Preferred
Recombinant Human Renin receptor (ATP6AP2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ATP6AP2. Target Synonyms: ATPase H(+)-transporting lysosomal accessory protein 2ATPase H(+)-transporting lysosomal-interacting protein 2ER-localized type I transmembrane adaptorEmbryonic liver differentiation factor 10N14FRenin/prorenin receptorVacuolar ATP synthase membrane sector-associated protein M8-9. Accession Number: O75787. Expression Region: 17~350aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 41.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4538