Learn More
Biomatik Corporation Recombinant Pig Complement C5a anaphylatoxin (C5), 100UG
Supplier:  Biomatik Corporation RPC24540
Encompass_Preferred
Recombinant Pig Complement C5a anaphylatoxin (C5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Pig (Sus scrofa; Porcine). Target Name: C5. Target Synonyms: C5; Complement C5a anaphylatoxin. Accession Number: P01032. Expression Region: 1~74aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 10.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5339