Learn More
Biomatik Corporation Recombinant Mouse T-cell surface glycoprotein CD3 epsilon chain (Cd3e), partial, 100UG
Supplier:  Biomatik Corporation RPC24566
Encompass_Preferred
Recombinant Mouse T-cell surface glycoprotein CD3 epsilon chain (Cd3e), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Cd3e. Target Synonyms: T-cell surface antigen T3/Leu-4 epsilon chainCD_antigen: CD3e. Accession Number: P22646. Expression Region: 23~108aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 11.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5342