Learn More
Biomatik Corporation Recombinant Human CD59 glycoprotein (CD59), 100UG
Supplier:  Biomatik Corporation RPC24572
Encompass_Preferred
Recombinant Human CD59 glycoprotein (CD59) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CD59. Target Synonyms: 1F5 antigen20 kDa homologous restriction factor; HRF-20; HRF20MAC-inhibitory protein; MAC-IPMEM43 antigen; Membrane attack complex inhibition factor; MACIFMembrane inhibitor of reactive lysis; MIRLProtectin; CD59. Accession Number: P13987. Expression Region: 26~102aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 11kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5343