Learn More
Biomatik Corporation Recombinant Human Collagen alpha-1 (XVIII) chain (COL18A1), partial, 100UG
Supplier:  Biomatik Corporation RPC24600
Encompass_Preferred
Recombinant Human Collagen alpha-1 (XVIII) chain (COL18A1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: COL18A1. Target Synonyms: Alpha 1 collagen type 18 (XVIII; COL18A1); Alpha 1 type XVIII collagen; Antiangiogenic agent; COIA1_HUMAN; COL15A1; Col18a1; Collagen alpha 1(XV) chain; Collagen alpha 1(XVIII) chain; Collagen alpha-1(XV) chain. Accession Number: P39060. Expression Region: 1578~1754aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 21.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5346